missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyl bromide, 98%, Thermo Scientific Chemicals

Product Code. 10305890
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-43-9
Molecular Formula C5H9Br
Molecular Weight (g/mol) 149.03
MDL Number MFCD00001359
InChI Key BRTFVKHPEHKBQF-UHFFFAOYSA-N
Synonym cyclopentyl bromide, cyclopentylbromide, cyclopentane, bromo, bromo cyclopentane, bromo-cyclopentane, bromocyclopentan, cylopentylbromide, cyclopentyl-bromide, sfphabilimup@, 4-bromocyclopentane
PubChem CID 8728
IUPAC Name bromocyclopentane
SMILES C1CCC(C1)Br

Specifications

Color Yellow to Brown
Density 1.3900g/mL
Boiling Point 137°C to 139°C
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C5H9Br
Refractive Index 1.4881 to 1.4901
Quantity 250 mL
Beilstein 05, 19
Specific Gravity 1.39
Solubility Information Solubility in water: insoluble in water
Formula Weight 149.03
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Cyclopentyl bromide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Cyclopentyl bromide
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.