missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentylacetyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11393586
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Cyclopentylacetyl chloride is used as a pharmaceutical intermediate and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentylacetyl chloride is used as a pharmaceutical intermediate and in chemical research.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Incompatible with bases and oxidizing agents. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1122-99-2
Molecular Formula C7H11ClO
Molecular Weight (g/mol) 146.61
MDL Number MFCD00236031
InChI Key NILLIUYSJFTTRH-UHFFFAOYSA-N
Synonym cyclopentylacetyl chloride, cyclopentaneacetyl chloride, cyclopentylacetylchloride, cylopentylacetyl chloride, acmc-1brgh, cyclopentaneacetylchloride, cyclopentylacetic chloride, cyclopentanacetyl chloride, cyclopentyl acetyl chloride, cyclopentyl-acetyl chloride
PubChem CID 3485182
IUPAC Name 2-cyclopentylacetyl chloride
SMILES ClC(=O)CC1CCCC1

Specifications

Density 1.087
Boiling Point 186°C
Flash Point 71°C (160°F)
Odor Pungent
Refractive Index 1.4625
Quantity 5 g
UN Number UN3265
Beilstein 1906113
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 146.62
Percent Purity 97%
Chemical Name or Material Cyclopentylacetyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.