missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentylmagnesium chloride, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

65.20 € - 378.00 €

Chemical Identifiers

CAS 32916-51-1, 60-29-7
Molecular Formula C5H9ClMg
Molecular Weight (g/mol) 128.88
MDL Number MFCD00001357
InChI Key GROGEABZCKQTJW-UHFFFAOYSA-M
Synonym cyclopentylmagnesium chloride, cyclopentyl magnesium chloride, cyclopentylmagnesium chloride solution, cylopentylmagnesium chloride, cyclopentylmagnesium cloride, magnesium,chlorocyclopentyl, cylopentyl magnesium chloride, cyclopentyl-magnesium chloride, grogeabzckqtjw-uhfffaoysa-m, chloro cyclopentyl magnesium
PubChem CID 3599845
IUPAC Name magnesium;cyclopentane;chloride
SMILES Cl[Mg]C1CCCC1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10527054
View Documents
Thermo Scientific Acros
208461000
100 mL AcroSeal™ Glass Bottle
65.20 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10071734
View Documents
Thermo Scientific Acros
208468000
800 mL AcroSeal™ Glass bottle
378.00 €
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

32916-51-1, 60-29-7
128.88
GROGEABZCKQTJW-UHFFFAOYSA-M
3599845
Cl[Mg]C1CCCC1
C5H9ClMg
MFCD00001357
cyclopentylmagnesium chloride, cyclopentyl magnesium chloride, cyclopentylmagnesium chloride solution, cylopentylmagnesium chloride, cyclopentylmagnesium cloride, magnesium,chlorocyclopentyl, cylopentyl magnesium chloride, cyclopentyl-magnesium chloride, grogeabzckqtjw-uhfffaoysa-m, chloro cyclopentyl magnesium
magnesium;cyclopentane;chloride

Specifications

32916-51-1, 60-29-7
−40°C
C5H9MgCl
0.86
GROGEABZCKQTJW-UHFFFAOYSA-M
magnesium;cyclopentane;chloride
3599845
Cyclopentylmagnesium chloride, 2M solution in diethyl ether
0.8600g/mL
C5H9ClMg
MFCD00001357
cyclopentylmagnesium chloride, cyclopentyl magnesium chloride, cyclopentylmagnesium chloride solution, cylopentylmagnesium chloride, cyclopentylmagnesium cloride, magnesium,chlorocyclopentyl, cylopentyl magnesium chloride, cyclopentyl-magnesium chloride, grogeabzckqtjw-uhfffaoysa-m, chloro cyclopentyl magnesium
Cl[Mg]C1CCCC1
128.88
128.88
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
In contact with water releases flammable gas.
Extremely flammable liquid and vapor.
May form explosive

GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

Product Identifier
  • Cyclopentylmagnesium chloride, 2M solution in diethyl ether

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 2

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H261-In contact with water releases flammable gases.
  • H224-Extremely flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.