missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Cyclophosphamide monohydrate, 97+%

Product Code. 11362457
missing translation for 'orderingAttributeHoverText'
Quantity:
1g
This item is not returnable. View return policy
This item is not returnable. View return policy
Applications
Cyclophosphamide monohydrate is used to treat cancers, autoimmune disorders and AL amyloidosis. It is a nitrogen mustard alkylating agent and reacts with deoxyribonucleic acid (DNA) to get its alkylated derivative. It is also involved in cancer research as a tool that induces apoptosis in tumor cells.

Solubility
Soluble in water. Slightly soluble ethanol, benzene, ethylene glycol, carbon tetrachloride, dioxane, ether, acetone, alcohol and chloroform.

Notes
Special handling precautions required. View MSDS prior to purchase, available online at www.alfa.comStore in cool place. Moisture and light senstive. Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Specifications

CAS 6055-19-2
Flash Point 113°C (235°F)
Odor Odorless
Molecular Formula C7H17Cl2N2O3P
MDL Number MFCD00149395
Quantity 1g
UN Number UN3464
Beilstein 8167897
Merck Index 14,2747
Synonym cyclophosphamide monohydrate, cyclophosphamide hydrate, cytoxan, endoxon, endoxan monohydrate, ciclophosphamide hydrat, cytoxan hydrate, endoxan a, cyclophosphamide hydrated
InChI Key PWOQRKCAHTVFLB-UHFFFAOYNA-N
SMILES O.ClCCN(CCCl)P1(=O)NCCCO1
IUPAC Name N,N-bis(2-chloroethyl)-2-oxo-1,3,2$l^{5}-oxazaphosphinan-2-amine;hydrate
Molecular Weight (g/mol) 279.10
PubChem CID 22420
ChEBI CHEBI:4026
Formula Weight 279.10 (261.09 Anhydrous)
Percent Purity ≥97%
Chemical Name or Material Cyclophosphamide monohydrate
Show More Show Less
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado