missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopropanemethylamine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Cyclopropanemethylamine used in studies of potential inhibitors of glycine cleavage system of brain and liver mitochondria. It is used in chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cyclopropanemethylamine used in studies of potential inhibitors of glycine cleavage system of brain and liver mitochondria. It is used in chemical synthesis.
Solubility
Fully miscible in water.
Notes
Air sensitive. Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, air , carbon dioxide.
Cyclopropanemethylamine used in studies of potential inhibitors of glycine cleavage system of brain and liver mitochondria. It is used in chemical synthesis.
Solubility
Fully miscible in water.
Notes
Air sensitive. Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, air , carbon dioxide.
Specifications
Specifications
Density | 0.82 |
Boiling Point | 85°C to 86°C |
Flash Point | 2°C (36°F) |
Odor | Amine-like |
Refractive Index | 1.433 |
Quantity | 5 g |
UN Number | UN2733 |
Beilstein | 773674 |
Sensitivity | Air Sensitive |
Solubility Information | Fully miscible in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'