Learn More
Cyclopropanesulfonyl chloride, 97%, Thermo Scientific Chemicals
290.45 € - 818.00 €
Chemical Identifiers
CAS | 139631-62-2 |
---|---|
Molecular Formula | C3H5ClO2S |
Molecular Weight (g/mol) | 140.58 |
MDL Number | MFCD01631933 |
InChI Key | PFWWSGFPICCWGU-UHFFFAOYSA-N |
Synonym | cyclopropanesulfonylchloride, chlorocyclopropylsulfone, cyclopropanesulphonyl chloride, cyclopropylsulfonyl chloride, cyclopropylsulphonyl chloride, cyclopropane sulfonyl chloride, cyclopropanesulfonic acid chloride, pubchem10869, acmc-209cje, cyclopropylsulfonylchloride |
PubChem CID | 2760919 |
IUPAC Name | cyclopropanesulfonyl chloride |
SMILES | ClS(=O)(=O)C1CC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11984581
|
Thermo Scientific Acros
439020010 |
1 g | Glass bottle |
290.45 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11994581
|
Thermo Scientific Acros
439020050 |
5 g | Glass bottle |
818.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
139631-62-2 | |
140.58 | |
PFWWSGFPICCWGU-UHFFFAOYSA-N | |
2760919 | |
ClS(=O)(=O)C1CC1 |
C3H5ClO2S | |
MFCD01631933 | |
cyclopropanesulfonylchloride, chlorocyclopropylsulfone, cyclopropanesulphonyl chloride, cyclopropylsulfonyl chloride, cyclopropylsulphonyl chloride, cyclopropane sulfonyl chloride, cyclopropanesulfonic acid chloride, pubchem10869, acmc-209cje, cyclopropylsulfonylchloride | |
cyclopropanesulfonyl chloride |
Specifications
139631-62-2 | |
1.3800g/mL | |
96°C | |
96% min. (GC) | |
MFCD01631933 | |
cyclopropanesulfonylchloride, chlorocyclopropylsulfone, cyclopropanesulphonyl chloride, cyclopropylsulfonyl chloride, cyclopropylsulphonyl chloride, cyclopropane sulfonyl chloride, cyclopropanesulfonic acid chloride, pubchem10869, acmc-209cje, cyclopropylsulfonylchloride | |
ClS(=O)(=O)C1CC1 | |
140.58 | |
140.59 | |
Liquid |
Colorless to Yellow | |
60.0°C (2.0 Torr) | |
Authentic | |
C3H5ClO2S | |
1.38 | |
PFWWSGFPICCWGU-UHFFFAOYSA-N | |
cyclopropanesulfonyl chloride | |
2760919 | |
97% | |
Cyclopropanesulfonyl chloride |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
GHS Signal Word: Danger
- Cyclopropanesulfonyl chloride, 97%
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only