missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanethiocarboxamide, 97%, Thermo Scientific Chemicals

Product Code. 15475027
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Cyclopropanecarbothioamide is used for purification in single-step preparation of thiazolo [5, 4-b] pyridine-and thiazolo [5, 4-c] pyridine derivatives from chloronitropyridines and thioamides, or thioureas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropanecarbothioamide is used for purification in single-step preparation of thiazolo [5, 4-b] pyridine-and thiazolo [5, 4-c] pyridine derivatives from chloronitropyridines and thioamides, or thioureas.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20295-34-5
Molecular Formula C4H7NS
Molecular Weight (g/mol) 101.167
MDL Number MFCD09469287
InChI Key IIPJWNFOLPDTEQ-UHFFFAOYSA-N
Synonym cyclopropanethiocarboxamide, cyclopropanecarbothioic acid amide, cyclopropanethioamide, cyclopropane carbothioamide, cyclopropane-thiocarboxamide, cyclopropane-1-carbothioamide, aminocyclopropylmethane-1-thione
PubChem CID 22140884
IUPAC Name cyclopropanecarbothioamide
SMILES C1CC1C(=S)N

Specifications

Melting Point 111°C to 113°C
Quantity 1 g
Formula Weight 101.17
Percent Purity 97%
Chemical Name or Material Cyclopropanethiocarboxamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.