missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylboronic acid, tech. 85%, Thermo Scientific Chemicals

Product Code. 11376719
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 411235-57-9
Molecular Formula C3H7BO2
Molecular Weight (g/mol) 85.90
MDL Number MFCD04038750
InChI Key WLVKDFJTYKELLQ-UHFFFAOYSA-N
Synonym cyclopropyl boronic acid, cyclopropylboranediol, cyclopropaneboronic acid, boronic acid, cyclopropyl, cyclopropyl-boronic acid, boronic acid, cyclopropyl-9ci, cyclopropylboronicacid, boronocyclopropane, cyclopropyboronic acid, cyclopropylboromc acid
PubChem CID 2760897
IUPAC Name cyclopropylboronic acid
SMILES OB(O)C1CC1

Specifications

Melting Point 90°C to 95°C
Quantity 1 g
Formula Weight 85.9
Percent Purity 85%
Grade Technical
Chemical Name or Material Cyclopropylboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.