missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Product Code. 11366759
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It often acts as the starting nutrient mix for dropout studies where fermentation conditions required the absence of one or more essential amino acids. For classifying Yeasts based on colonial characteristics and cell morphology.

Solubility
Soluble in water (17 mg/ml), (25°C). Soluble in organic solvents.

Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, water and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23719-80-4
Molecular Formula C3H5BrMg
Molecular Weight (g/mol) 145.28
MDL Number MFCD00672094
InChI Key QUPLLJGAJVXXBW-UHFFFAOYSA-M
Synonym cyclopropylmagnesium bromide, cyclopropylmagnesiumbromide, bromo cyclopropyl magnesium, magnesium, bromocyclopropyl, cyclopropyl magnesium bromide, cprmgbr, cyclopropyl magnesiumbromide, magnesium,bromocyclopropyl, bromo-cyclopropyl magnesium, cyclopropylmagnesiunn bromide, grignard reagent
PubChem CID 11116189
IUPAC Name magnesium;cyclopropane;bromide
SMILES Br[Mg]C1CC1

Specifications

Density 0.968
Flash Point −17°C (1°F)
Quantity 25 mL
UN Number UN2924
Sensitivity Air and moisture sensitive; May form a precipitate
Solubility Information Soluble in water (17mg/ml),(25°C). Soluble in organic solvents.
Formula Weight 145.28
Concentration or Composition (by Analyte or Components) 0.5 to 0.7M in THF
Chemical Name or Material Cyclopropylmagnesium bromide
compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.