Learn More
Cytiva CyDye™ DIGE Fluor Minimal Dye Labeling Kits
Allow detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel. Amersham CyDye™ DIGE Fluor Minimal Labeling Kits contain size- and charge-matched fluorescent dyes specifically designed for detecting protein abundance differences in 2-D Fluorescence Difference Gel Electrophoresis using Ettan DIGE system.
Brand:  Cytiva 28-9345-30
 View more versions of this product
Product Code. 10066084
  • 1040.00 € / 1 set
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Size- and charge-matched dyes enable co-migration of labeled samples within the gel
  • Bright and highly sensitive dyes allow the use of the minimal labeling technique
  • Minimal loss of signal during labeling, separation, and scanning
  • No change in signal over wide pH range used during first-dimension (IEF) separation
  • Discrete signal from each fluor with minimal cross-talk contributes to high accuracy
All minimal dyes allow the labeling of 50μg protein per labeling reaction. Minimal Dye Labeling Kits contain 2nmol or 5nmol of each of the minimal dyes.
For general 2-D applications where sufficient amounts of sample are available.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Solid
2 nmol of each: Cy2, Cy3, and Cy5
2nM of CyDye DIGE Fluor minimal labeling kit (Cy2, Cy3 and Cy5)
Blue, Green, Red
The Scarce Sample Labeling Kit is designed to be used for applications in which only small amounts of sample are available, for example, in laser capture microdissection, detects up to 3 prelabeled protein samples, standard on the same 2-D electrophoresis gel
Amersham CyDye DIGE Fluor Minimal Dye Labeling Kit
-15°C to -30°C
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Cy2, Cy3, Cy5