missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva CyDye™ DIGE Fluors: Cy2
Allow detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel
989.00 € - 3268.00 €
Specifications
Format | Solid |
---|---|
For Use With (Application) | For general 2-D applications where sufficient amounts of sample are available, detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel |
Product Type | Amersham CyDye DIGE Fluor Cy2 Minimal Dye |
Content And Storage | -15°C to -30°C |
Color | Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10075624
|
Cytiva
RPK0272 |
5 x 5 nM |
3268.00 €
Each |
Estimated Shipment: 14-03-2025
Log in to see stock. |
|||||
10648285
|
Cytiva
25-8010-82 |
5 nM |
989.00 €
Each |
In Stock
Log in to see stock. |
|||||
10156904
|
Cytiva
25-8008-60 |
2 x 5 nM | N/A | ||||||
Description
- Size- and charge-matched dyes enable co-migration of labeled samples within the gel
- Bright and highly sensitive dyes allow the use of the minimal labeling technique
- Minimal loss of signal during labeling, separation, and scanning
- No change in signal over wide pH range used during first-dimension (IEF) separation
- Discrete signal from each fluor with minimal cross-talk contributes to high accuracy
- Allow the labeling of 50μg protein per labeling reaction
For general 2-D applications where sufficient amounts of sample are available.
Specifications
Solid | |
Amersham CyDye DIGE Fluor Cy2 Minimal Dye | |
Blue |
For general 2-D applications where sufficient amounts of sample are available, detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel | |
-15°C to -30°C |
Safety and Handling
Signal Word
Hazard Category
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title