missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva CyDye™ DIGE Fluors: Cy5
Allow detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel
989.00 € - 3268.00 €
Specifications
Format | Solid |
---|---|
For Use With (Application) | For general 2-D applications where sufficient amounts of sample are available, detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel |
Product Type | Amersham CyDye DIGE Fluor Cy5 Minimal Dye |
Content And Storage | -15°C to -30°C |
Color | Red |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10269114
|
Cytiva
RPK0275 |
5 x 5 nM |
3268.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||||
10116624
|
Cytiva
25-8008-62 |
2 x 5 nM | N/A | ||||||
10407104
|
Cytiva
25-8010-85 |
5 nM |
989.00 €
Each |
In Stock
Log in to see stock. |
|||||
Description
- Size- and charge-matched dyes enable co-migration of labeled samples within the gel
- Bright and highly sensitive dyes allow the use of the minimal labeling technique
- Minimal loss of signal during labeling, separation, and scanning
- No change in signal over wide pH range used during first-dimension (IEF) separation
- Discrete signal from each fluor with minimal cross-talk contributes to high accuracy
- Allow the labeling of 50μg protein per labeling reaction
For general 2-D applications where sufficient amounts of sample are available.
Specifications
Solid | |
Amersham CyDye DIGE Fluor Cy5 Minimal Dye | |
Red |
For general 2-D applications where sufficient amounts of sample are available, detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel | |
-15°C to -30°C |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Acute toxicity Category 4
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H317-May cause an allergic skin reaction.
- H332-Harmful if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501-Dispose of contents/container to .