Learn More
Thermo Scientific Chemicals Cylindrical Al-23 Crucibles
Brand: Thermo Scientific Chemicals 032953.KF
106.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
30mm | |
1 Piece | |
35mm | |
Cylindrical Al-23 Crucibles |
Al-23 | |
40 mL | |
50mm |
Product Suggestions
Customers who viewed this item also viewed.