Learn More
Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels, with Glass Plug
For the safe, easy and accurate transfer of fluids in the laboratory. Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels, with Glass Plug are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 04153711
 View more versions of this product
Product Code. 10303291
  • 135.00 € / Pack of 2
Estimated Shipment: 20-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With NS socket and cone
  • With scale (approx. volume/mL)
  • With inscription label
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
100 mL
14/23 NS
Yes
Borosilicate Glass
Without Pressure Equalizing Tube
Dropping Funnel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels, with Glass Plug > Capacity: 100mL; Socket: 14/23 NS; Cone 14/23 NS