Learn More
Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels, with Glass Plug
For the safe, easy and accurate transfer of fluids in the laboratory. Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels, with Glass Plug are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 04152811
 View more versions of this product
Product Code. 10644661
  • 125.00 € / Pack of 2
Estimated Shipment: 28-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With NS socket and cone
  • With scale (approx. volume/mL)
  • With inscription label
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
50 mL
14/23 NS
Yes
Borosilicate Glass
Without Pressure Equalizing Tube
Dropping Funnel