missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Pressure Equalizing Dropping Funnels, with PTFE Plug

Product Code. 10707711
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Joint Number:
14/23 NS
29/32 NS
Includes:
Pressure Equalizing Tube
w/o Pressure Equalizing Tube
Unit Size:
Each
Pack of 2
This item is not returnable. View return policy

Product Code. 10707711

Brand: Lenz Laborglasinstrumente™ 04372211

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Useful for transferring fluids and handling air-sensitive reagents in a sealed, inert-gas environment. Lenz Laborglasinstrumente™ Pressure Equalizing Dropping Funnels are manufactured from durable DURAN™ borosilicate glass.

  • With scale (approx. volume/mL)
  • With pressure equalizing tube
  • With screwthread retaining device
  • Cylindrical shape with NS socket and cone
  • Corrosion-free and resistant to thermal shock

Specifications

Capacity (Metric) 25 mL
Material Borosilicate Glass
Graduated Yes
Includes Pressure Equalizing Tube
Joint Number 14/23 NS
Product Type Dropping Funnel
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.