Learn More
Fisherbrand™ Cylindrical PTFE or Glass Stir Bars
Alnico V magnet core encapsulated by PTFE or glass . Fisherbrand™ Cylindrical PTFE or Glass Stir Bars are autoclavable.
Brand:  Fisherbrand™ 001.210.6
 View more versions of this product
Product Code. 11878932
  • 18.05 € / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Stable, general-purpose magnetic stir bar
  • Smooth, round profile for a wide variety of applications
  • Autoclavable
Specifications
Specifications
Autoclavable
White
Containers & Vessels / Stirrer Hot Plates
6 mm
10/Pk.
PTFE Stir Bars Cylindrical
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
PTFE encapsulated Alnico V magnet core
PTFE
10 mm
Cylinder