Learn More
Fisherbrand™ PTFE-Coated Rare Earth Core Stir Bars, Cylindrical
Use for significantly increased magnetic strength and resistance to demagnetization. Rare earth stir bars are easily identified by a chemically inert carbon black spot.
Brand:  Fisherbrand™ 001.130.RE
 View more versions of this product
Product Code. 16205741
  • 22.10 € / Pack of 5
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Use rare earth magnets for significantly increased magnetic strength compared to those using AlNiCo magnets
  • Samarium-cobalt core is resistant to demagnetization
  • Identified by a chemically inert, carbon black spot
The high magnetic coupling power of rare earth stir bars may cause abrasion of the PTFE coating against the base of the container. When stirring arrays are used, as in multi-well plates, lateral interaction between stir bars may be observed.
Specifications
Specifications
Autoclavable
White
Containers & Vessels / Stirrer Hot Plates
30 mm
Cylinder
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
PTFE encapsulated Samarium-Cobalt magnet core
6 mm
2/Pk.