missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ PTFE-Coated Rare Earth Core Stir Bars, Cylindrical
Use for significantly increased magnetic strength and resistance to demagnetization. Rare earth stir bars are easily identified by a chemically inert carbon black spot.
Brand: Fisherbrand™ 001.112.RE
Description
- Use rare earth magnets for significantly increased magnetic strength compared to those using AlNiCo magnets
- Samarium-cobalt core is resistant to demagnetization
- Identified by a chemically inert, carbon black spot
Specifications
Autoclavable | |
White | |
Containers & Vessels / Stirrer Hot Plates | |
12 mm | |
Cylinder |
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) | |
PTFE encapsulated Samarium-Cobalt magnet core | |
4.5 mm | |
2/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction