Learn More
Lenz Laborglasinstrumente™ Cylindrical Reaction Vessels with Thermostatic Jacket
Excellent for containing reactants and suitable for vacuum purposes. Lenz Laborglasinstrumente™ Cylindrical Reaction Vessels with Thermostatic Jacket are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 06011588
 View more versions of this product
Product Code. 11542264
  • 3905.00 € / Each
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With graduation and laboratory flange LF
  • With flat bottom and two connectors (with threads GL 14/18 or ball flanges KF 15/25)
  • Delivery with two plastic olives (GL 14/18) and screw caps
  • Corrosion-free and resistant to thermal shock
  • Not suitable for the use under positive pressure
Specifications
Specifications
DURAN™ Tubing
Borosilicate Glass, Duran Glass
Laboratory Flange 200
Cylindrical
250 mm
10 L
495 mm
Reaction vessel
Flat