missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cynomolgus Monkey MuSK (XP_005581150.1) VersaClone cDNA

Product Code. 18783967
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18783967

Brand: R&D Systems™ RDC3538

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Gene MuSK
Accession Number XP_005581150.1
Gene ID (Entrez) 4593
Gene Alias EC 2.7.10, EC 2.7.10.1, MGC126323, MGC126324, muscle, skeletal, receptor tyrosine kinase, MuSK, skeletal receptor tyrosine-protein kinase
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Cynomolgus Monkey
Quantity 10 μg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.