Learn More
CYP20A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16702-1-AP-150UL
 View more versions of this product
Product Code. 16826424
  • 410.00 € / 150µL
Explore more special offers
Description
Description
CYP20A1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases that catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. The protein lacks one amino acid of the conserved heme binding site. It also lacks the conserved I-helix motif AGX(D, E)T, suggesting that its substrate may carry its own oxygen.
Specifications
Specifications
CYP20A1
Polyclonal
Unconjugated
CYP20A1
CYP M, CYP20A1, Cytochrome P450 20A1
Rabbit
Antigen Affinity Chromatography
RUO
316435, 57404, 77951
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6P7D4, Q6UW02, Q8BKE6
Cyp20a1
CYP20A1 Fusion Protein Ag10099
150 μL
Primary
Human, Mouse, Rat