Learn More
Thermo Scientific™ CYP2C8 BACULOSOMES™ Plus Reagent, rHuman
Cytochrome P450 BACULOSOMES™ Plus Reagents are microsomes prepared from insect cells infected with recombinant baculovirus containing a human CYP450 isozyme, as well as human cytochrome P450 reductase.
Brand: Thermo Scientific™ PV6138
Description
Cytochrome P450 BACULOSOMES™ Plus Reagents are microsomes prepared from insect cells infected with recombinant baculovirus containing a human CYP450 isozyme, as well as human cytochrome P450 reductase. For this particular isozyme (CYP2C8), human cytochrome b5 is also included.
- Single human P450 isozyme for detailed drug metabolism studies
- High specific activity increases dynamic range and sensitivity
- No background metabolism for clear results
Single Overexpressed Human CYP450 Isozyme
CYP450 BACULOSOMES™ Plus Reagents offer a distinct advantage over Human Liver Microsomes (HLMs) in that only one CYP450 isozyme is expressed, thereby preventing metabolism by other CYP450s or other classes of drug metabolizing enzymes.
High Activity Results in Increased Sensitivity
Compound rankings based on metabolism and inhibition profiles observed with CYP450 BACULOSOMES™ Plus Reagents are very similar to those seen with HLMs for most compounds tested. However, CYP450 BACULOSOMES™ Plus Reagents activity and metabolic rates with most substrates is significantly higher than those seen with HLMs. This results in a broad dynamic range and high sensitivity in assays utilizing CYP450 BACULOSOMES™ Plus Reagents. The high level of activity and reproducibility of the enzyme component observed in the reaction with most probe substrates makes CYP450 BACULOSOMES™ Plus Reagents well suited to high-throughput screening formats.
High Signal to Noise Ratio
No background metabolism by endogenous insect CYP450s has been detected with any of the Vivid™ probe substrates tested so far.
Applications:
- Isozyme identification
- DMPK analysis
- Isozyme-specific metabolism
- Inhibition screening

Specifications
CYP2C8 BACULOSOMES™ Plus Reagent, 0.5nmol, Liquid, 4.6mg/mL Concentration, Cytochrome P450 Druggable Target, Microsomes Sub-Cellular Localization, Cytochrome P450 Druggable Target, Baculovirus Expression, Human Species, Used for Cell Viability, Proliferation and Function, Pharma and Biopharma | |
Microplate Reader | |
Cytochrome P450 | |
Cell viability, proliferation and function, hepatocytes and hepatic models, pharma and biopharma, protein assays and analysis, target-based ADME/Tox assays | |
1 tube, store at -80°C. | |
0.5 nmol | |
Cytochrome P450 Enzymes | |
Microsomes | |
Human |
4.6mg/mL | |
BACULOSOMES™ | |
Liquid | |
0.5nmol | |
Bacterial Microsome | |
Baculovirus | |
Dry Ice | |
Toxicology & Drug Metabolism, Drug Development |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2911323 | Certificate of Analysis | 21/02/2025 | PV6138 |
2611443 | Certificate of Analysis | 04/05/2023 | PV6138 |
2380752 (1) | Certificate of Analysis | 30/09/2022 | PV6138 |
2041598 | Certificate of Analysis | 24/01/2019 | PV6138 |
1883451 | Certificate of Analysis | 19/07/2017 | PV6138 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.