Learn More
Cyproterone acetate, 98%, Thermo Scientific Chemicals
67.20 € - 225.00 €
Chemical Identifiers
CAS | 427-51-0 |
---|---|
Molecular Formula | C24H29ClO4 |
Molecular Weight (g/mol) | 416.94 |
InChI Key | UWFYSQMTEOIJJG-FDTZYFLXSA-N |
Synonym | cyproterone acetate, androcur, cyproterone 17-o-acetate, cyproteron acetate, cyproteron-r acetate, cyprosterone acetate, unii-4km2bn5jhf, cyproteroneacetate, cyproterone 17alpha-acetate, ccris 4385 |
PubChem CID | 9880 |
ChEBI | CHEBI:50743 |
SMILES | CC(=O)C1(CCC2C1(CCC3C2C=C(C4=CC(=O)C5CC5C34C)Cl)C)OC(=O)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15549483
|
Thermo Scientific Acros
459542500 |
250 mg |
67.20 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15595286
|
Thermo Scientific Acros
459540010 |
1 g |
225.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
427-51-0 | |
416.94 | |
cyproterone acetate, androcur, cyproterone 17-o-acetate, cyproteron acetate, cyproteron-r acetate, cyprosterone acetate, unii-4km2bn5jhf, cyproteroneacetate, cyproterone 17alpha-acetate, ccris 4385 | |
CHEBI:50743 |
C24H29ClO4 | |
UWFYSQMTEOIJJG-FDTZYFLXSA-N | |
9880 | |
CC(=O)C1(CCC2C1(CCC3C2C=C(C4=CC(=O)C5CC5C34C)Cl)C)OC(=O)C |
Specifications
427-51-0 | |
0.1% max. | |
Authentic | |
C24H29ClO4 | |
Solubility in water: insoluble in water. Other solubilities: soluble in acetone, dichloromethane, methanol, slightly soluble in ethanol | |
CC(=O)C1(CCC2C1(CCC3C2C=C(C4=CC(=O)C5CC5C34C)Cl)C)OC(=O)C | |
9880 | |
416.94 | |
Cyproterone acetate |
207.0°C to 215.0°C | |
0.5% max. (80°C) | |
97.5% min. (HPLC) | |
cyproterone acetate, androcur, cyproterone 17-o-acetate, cyproteron acetate, cyproteron-r acetate, cyprosterone acetate, unii-4km2bn5jhf, cyproteroneacetate, cyproterone 17alpha-acetate, ccris 4385 | |
UWFYSQMTEOIJJG-FDTZYFLXSA-N | |
416.94 | |
CHEBI:50743 | |
98% |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
Suspected of causing cancer.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 207-048-3
- Cyproterone acetate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
Hazard Statement
- H351-Suspected of causing cancer.
- H332-Harmful if inhaled.
- H312-Harmful in contact with skin.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only