missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ CyQUANT™ NF Cell Proliferation Assay
CyQUANT™ NF Cell Proliferation Assay Kit
300.00 € - 676.00 €
Specifications
Content And Storage | Store in refrigerator 2°C to 8°C and protect from light. |
---|---|
Detection Method | Fluorescence |
For Use With (Application) | Proliferation Assay |
For Use With (Equipment) | Microplate Reader |
Product Type | Cell Proliferation Assay |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10247072
|
Molecular Probes™
C35006 |
1,000 assays |
676.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10709253
|
Molecular Probes™
C35007 |
200 assays kit |
300.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CyQUANT™ NF Cell Proliferation Assay Kit provides a fast and sensitive method for counting cells in a population and measuring proliferation in microplate format.Features of the CyQUANT™ NF Cell Proliferation Assay Kit:
A Rapid and simple method for measuring cell proliferation
The CyQUANT™ NF Cell Proliferation assay does not require cell lysis, long incubations, radioactivity, or removal of stain from cells. The CyQUANT™ NF assay eliminates the freeze-thaw cell lysis step of the original CyQUANT™ cell proliferation assay by using a cell-permeant DNA-binding dye in combination with a plasma membrane permeabilization reagent. The CyQUANT™ NF Cell Proliferation Assay Kit can be used in either 96-well or 384-well microplate formats, and is available in two configurations: a 200 assay kit (C35007) for smaller sample sizes, and a 1000 assay kit (C35006) for high-throughput applications.
AlamarBlue™ is a registered trademark of TREK Diagnostic Systems.
Specifications
Store in refrigerator 2°C to 8°C and protect from light. | |
Proliferation Assay | |
Cell Proliferation Assay | |
384-well plate, 96-well plate | |
Room Temperature |
Fluorescence | |
Microplate Reader | |
Other Label(s) or Dye(s) | |
CyQUANT™ |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title