missing translation for 'onlineSavingsMsg'
Learn More
Cytarabine, 98%, Thermo Scientific Chemicals Product Code.: 15244774

Cytarabine, 98%, Thermo Scientific Chemicals

Product Code. 15244774
100 mg, Glass bottle
Click to view available options
Quantity:
100 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 15244774

Brand: Thermo Scientific Acros 449561000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cytarabine, CAS # 147-94-4, is an inhibitor of DNA synthesis that is used as an anticancer and antiviral agent.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Cytarabine is a nucleoside analog that acts as an antileukemic agent
  • Cytarabine becomes cytotoxic following conversion to its triphosphorylated form, which can be incorporated into DNA primers by DNA polymerase alpha
  • Can induce S phase cell cycle arrest and apoptosis in leukemic and neuronal cells

Applications

  • Has been used to study chemotherapeutic regimens in laboratory studies of leukemia
  • Useful for laboratory studies of cytarabine resistance mechanisms
  • At nanomolar concentrations, cytarabine has been seen to induce mitochondrial apoptosis in leukemic cells through upregulation of multiple Bcl-2 family members
  • It can induce neuronal cell death via inducing γ-H2AX phosphorylation in histones, in the absence of double-stranded DNA breaks
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147-94-4
Molecular Formula C9H13N3O5
Molecular Weight (g/mol) 243.22
InChI Key UHDGCWIWMRVCDJ-CCXZUQQUSA-N
Synonym cytarabine, cytosine arabinoside, ara-c, arabinocytidine, cytosar, arabinosylcytosine, aracytidine, cytarabin, cytarabinoside, tarabine
PubChem CID 6253
ChEBI CHEBI:28680
IUPAC Name 4-amino-1-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)O

Specifications

Melting Point 220.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Merck Index 152781
Solubility Information Solubility in water: soluble. Other solubilities: soluble in dmso, ethanol
Formula Weight 243.22
Percent Purity 98%
Chemical Name or Material Cytarabine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465608 Certificate TypeCertificate of Analysis Date26/08/2024 Catalog Number449560050,449560010,449561000
Lot NumberA0425805 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number449560050,449560010,449561000
Lot NumberA0444786 Certificate TypeCertificate of Analysis Date06/08/2022 Catalog Number449560050,449560010,449561000
Lot NumberA0374024 Certificate TypeCertificate of Analysis Date15/12/2020 Catalog Number449560050,449560010,449561000
Lot NumberA0333607 Certificate TypeCertificate of Analysis Date02/11/2020 Catalog Number449560050,449560010,449561000
compliance-icons compliance-icons
Product Identifier
  • Cytosine beta-D-arabinofuranoside
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H361d-Suspected of damaging the unborn child.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytarabine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.