Learn More
Thermo Scientific™ Cytochalasin A
Brand: Thermo Scientific™ J60280.#0
View more versions of this product
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
5 mg | |
2°C to 8°C | |
14110-64-6 | |
477.60 | |
191°C | |
Powder |
White | |
An anti-cytoskeletal compound. Inhibits glucose transport, actin polymerization and blocks the formation of microtubules. Inhibits cell divisiion. Inhibits HIV-1 protease | |
C29H35NO5 | |
Light Sensitive | |
Soluble in DMF, DMSO, ethanol and acetone. Insoluble in water. |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.