missing translation for 'onlineSavingsMsg'
Learn More

Cytochrome C From Horse Heart MP Biomedicals

Product Code. 11453430
Click to view available options
Quantity:
500 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 9007-43-6
Molecular Formula C40H48FeN6O6S4
Molecular Weight (g/mol) 892.94
MDL Number MFCD00130890
InChI Key UMUOLVQYJUIZKD-RHANTFMNSA-L
Synonym cytochrome c, 3-7,12-bis 1-2-amino-3-methylamino-3-oxopropyl sulfanylethyl-18-2-carboxyethyl-3,8,13,17-tetramethylporphyrin-21,23-diid-2-yl propanoic acid; iron 2+
PubChem CID 16057918
IUPAC Name 3-[8,13-bis[1-[2-amino-3-(methylamino)-3-oxopropyl]sulfanylethyl]-18-(2-carboxyethyl)-3,7,12,17-tetramethylporphyrin-21,24-diid-2-yl]propanoic acid;iron(2+)
SMILES CC(SC(=O)[C@@H](N)CS)C1=C(C)\C2=C\C3=C(C(C)SC(=O)[C@@H](N)CS)C(C)=C4CC5N=C(\C=C6/N([Fe++]N34)\C(=C/C1=N2)C(C)=C6CCC(O)=O)C(CCC(O)=O)=C5C

Specifications

Quantity 500 mg
Formula Weight 12000
Chemical Name or Material Cytochrome C, From Horse Heart
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.