missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome C From Horse Heart MP Biomedicals
Click to view available options
Quantity:
500 mg
1 g
Specifications
Specifications
Quantity | 1 g |
Formula Weight | 12000 |
Chemical Name or Material | Cytochrome C, From Horse Heart |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction