Learn More
Cytiva Cytodex 3 Gamma Microcarriers
The microcarrier of choice for cells that are difficult to culture in vitro - particularly for cells with an epithelial-like morphology.
Brand:  Cytiva 17548802
 View more versions of this product
Product Code. 15757207
  • 6250.00 € / 300g
Explore more special offers
Description
Description
  • Cytodex 3 Gamma is designed by chemically coupling of a thin layer of denatured porcine collagen type 1 to the cross-linked dextran matrix.
  • As the collagen surface layer can be digested by a variety of proteolytic enzymes, harvest of such cells from the microcarriers can be performed with maintained high cell viability and membrane integrity.
  • This feature of Cytodex 3 Gamma is beneficial in the development of successful serial subcultivation protocols for scaling up culture volumes.
  • Applications include the production of large quantities of cells, viruses and recombinant cell products (e.g. interferon, enzymes, nucleic acids, hormones), studies on cell adhesion, differentiation and cell function, perfusion column culture systems, microscopy studies, harvesting mitotic cells, isolation of cells, membrane studies, storage and transport of cells, assays involving cell transfer and studies on uptage of labelled compounds
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Production of large quantities of cells, viruses and recombinant cell products, studies on cell adhesion, differentiation and cell function
300 g