Learn More
CytoScint ES, MP Biomedicals™
Liquid Scintillation Cocktails; CytoScint ES; MP Biomedical; 4 x 1 gal.; Biodegradable
Brand:  MP Biomedicals 0188245301
 View more versions of this product
Product Code. 15702252
  • 353.00 € / 3.79 L
Explore more special offers
Specifications
Liquid Scintillation Cocktails
1 gal.
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • cytoscint ES
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : phenylxylylethane, linear alkylbenzene,ethoxylated nonylphenol, 2-methoxyethanol