Learn More
Epredia™ Cytospin™ 4 Cytocentrifuge Sealed Head Rotor
Use these accessories with the Epredia™ Cytospin™ 4 Cytocentrifuge.
114.00 € - 1400.00 €
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
12698026
|
Epredia™
59910019 |
Sealed Head Seals |
114.00 €
Pack of 3 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12688026
|
Epredia™
59910018 |
Sealed Head Rotor |
1400.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Shorten preparation time by having a second sealed head ready when the first finishes processing
- Autoclavable
- 12-chamber head seals completely