Learn More
Epredia™ Cytospin™ 4 Cytofunnel™ Starter Kit
Accessory Starter Kit for the Epredia™ Cytospin™ 4 Cytocentrifuge. (Centrifuge is sold separately, Cat. No. A78300003)
Brand: Epredia™ A78710013
816.90 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
The Cytofunnel Starter Kit provides everything needed for new Cytospin 4 users.
Available in both standard clipped Cytofunnel and EZ Cytofunnel options:
- Clipped Cytofunnel Starter Kit
- Includes: 50 Single White Cytofunnels, 25 Double Cytofunnels, Cytoslides, 12 Cytoclips
- EZ Cytofunnel Starter Kit
- Includes: 25 EZ Megafunnels, 20 EZ Single White Cytofunnels, 20 EZ Double Cytofunnels and Cytoslides
Specifications
Cytospin™ 4 Cytocentrifuge | |
EZ Cytofunnel Starter Kit | |
100/240 V |
Cytospin | |
50/60 Hz |