Learn More
Invitrogen™ CytoTune™-iPS 2.0 Sendai Reprogramming Kit
The CytoTune™-iPS 2.0 Sendai Reprogramming Kit improves upon the well-referenced CytoTune™-iPS Sendai Reprogramming Kit.
Brand:  Invitrogen™ A16518
 View more versions of this product
Product Code. 15303976
  • 7700.00 € / 3 sets
Explore more special offers
Description
Description
  • This system continues to be easy to use and works with many different somatic cell types, using Sendai particles to deliver Yamanaka factors, which have been shown to be critical for efficient generation of iPSCs. This kit includes three vector preparations: polycistronic Klf4–Oct3/4–Sox2, cMyc, and Klf4.
  • Ease of use - still requires only one application of the virus to obtain successful reprogramming
  • Increased efficiency - usually at least 2-fold higher reprogramming efficiencies than the original kit
  • Rapid clearance - use the non-integrating Sendai virus that remains in the cytoplasm, reducing the likelihood of integration into the host genome. With the CytoTune™-iPS 2.0 Sendai virus, the Sendai backbone and transgenes clear the cytoplasm faster than the original.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
3 x 3 Tubes, store (at -68°C to -85°C.)
Kit
CytoTune™
3 Pack
Dry Ice
Sendai virus
Stem Cell Engineering & Reprogramming
Sendai Reprogramming Kit
Proprietary
Cytoplasm