missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CytoTune™-iPS 2.0 Sendai Reprogramming Kit
The CytoTune™-iPS 2.0 Sendai Reprogramming Kit improves upon the well-referenced CytoTune™-iPS Sendai Reprogramming Kit.
3140.00 € - 7700.00 €
Specifications
Delivery Type | Sendai virus |
---|---|
Format | Kit |
For Use With (Application) | Stem Cell Engineering & Reprogramming |
Product Line | CytoTune™ |
Product Type | Sendai Reprogramming Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15286620
|
Invitrogen™
A16517 |
1 Pack |
3140.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15303976
|
Invitrogen™
A16518 |
3 Pack |
7700.00 €
3 sets |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- This system continues to be easy to use and works with many different somatic cell types, using Sendai particles to deliver Yamanaka factors, which have been shown to be critical for efficient generation of iPSCs. This kit includes three vector preparations: polycistronic Klf4–Oct3/4–Sox2, cMyc, and Klf4.
- Ease of use - still requires only one application of the virus to obtain successful reprogramming
- Increased efficiency - usually at least 2-fold higher reprogramming efficiencies than the original kit
- Rapid clearance - use the non-integrating Sendai virus that remains in the cytoplasm, reducing the likelihood of integration into the host genome. With the CytoTune™-iPS 2.0 Sendai virus, the Sendai backbone and transgenes clear the cytoplasm faster than the original.
Specifications
Sendai virus | |
Stem Cell Engineering & Reprogramming | |
Sendai Reprogramming Kit | |
Dry Ice |
Kit | |
CytoTune™ | |
Proprietary | |
Cytoplasm |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title