missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CytoVista™ 3D Cell Culture Clearing/Staining Kit

CytoVista™ 3D Cell Culture Clearing/Staining Kit clears fluorescently labeled 3D cultured cells such as organoids and spheroids. 

Brand:  Invitrogen™ V11325

Product Code. 15961322

  • 600.00 € / 1 set
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Clearing process enables sharp and bright 3D fluorescent imaging for depths up to 1000 μm. The kit includes CytoVista™ 3D Cell Culture Clearing Reagent, penetration buffer, wash buffer, blocking buffer, and antibody dilution buffer.

Features of the CytoVista™ family of tissue and 3D cell culture clearing reagents include:
• Rapid clearing of fluorescently labeled tissue for 3D imaging
• Minimal shrinkage, expansion, or other morphological changes to tissue
• Simple workflow does not require any special instruments or equipment and is easily adapted to IHC
• Compatible with most fluorophores, including dyes and fluorescent proteins

The CytoVista™ tissue and 3D cell culture clearing workflow clears tissue or 3D cultured cells with minimal changes to morphology or the sensitivity of detection with almost any fluorophore. With an easy-to-use protocol, whole mouse brain (8-mm section) can be cleared in 24 hours, or a 1-mm section in 2 hours, without the use of any special instruments or equipment.

The CytoVista™ clearing workflow is compatible with most fluorophores, including fluorescent proteins that are detected with common fluorescent imaging instruments, such as wide-field microscopes, high content analyzers, confocal microscopes, and lightsheet microscopes. For first time users, we recommend the CytoVista™ Tissue Clearing/Staining Kit (Cat No V11324) or CytoVista™ 3D Cell Culture Clearing/Staining Kit (Cat No. V11325). These kits contain all necessary reagents for both clearing and IHC staining.

CytoVista™ clearing technology sufficiently clears tissue for 3D fluorescent imaging, but unlike other clearing techniques does not significantly changes the tissue morphology. Minimal morphological changes, such as shrinkage and contraction, have been observed. For precious samples, the clearing can be reversed, and the tissue can be processed for histology studies, like H&E staining.

For tissues that are easier to clear, like brain, lung, intestine, skin, and tissues of less than 250-μm thickness, treatment with CytoVista™ Tissue Clearing Reagent is sufficient for 3D imaging. However, tissues of more than 250-μm thickness or that are hard to clear like kidney, liver, heart, and placenta will require an initial treatment with CytoVista™ Tissue Clearing Reagent followed by treatment with CytoVista™ Tissue Clearing Enhancer.

CytoVista™ 3D Cell Culture Clearing Reagent is recommended for spheroids or organoids up to 1 mm in thickness.

For hard tissues like bone, cartilage, and connective tissue, CytoVista™ Tissue Clearing Reagent is not recommended.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

3D Cell Culture Clearing/Staining Kit
Cell & Tissue Preparation Reagent
1 kit
CytoVista
  • CytoVista 3D Cell Culture Clearing/Staining Kit, store at room temperature
  • CytoVista Antibody Penetration Buffer, 30 mL, store at 2°C to 8°CC
  • CytoVista 10X Wash Buffer, 70 mL, store at 2°C to 8°CC
  • CytoVista Blocking Buffer, 30 mL, store at 2°C to 8°CC
  • CytoVista Antibody Dilution Buffer, 30 mL, store at 2°C to 8°CC

    Due to variable shipping temperatures, content can be shipped in mulitple containers.
Approved for shipment at Room Temperature or on Wet Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.