missing translation for 'onlineSavingsMsg'
Learn More

D-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid, 98%, Thermo Scientific™

Product Code. 11936511
Click to view available options
Quantity:
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 103733-65-9
Melting Point 304°C to 306°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C10H11NO2
MDL Number MFCD00144038
Quantity 500 mg
Specific Rotation 156.5
Synonym r-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, 3r-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, h-d-tic-oh, d-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, r-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, r-+-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 3-isoquinolinecarboxylic acid, 1,2,3,4-tetrahydro-, 3r, 3r-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, 3r-1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid, r-1,2,3,4-tetrahydro-3-isoquinolinecarboxylicacid
InChI Key BWKMGYQJPOAASG-UHFFFAOYNA-N
SMILES OC(=O)C1CC2=CC=CC=C2CN1
IUPAC Name (3R)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
Molecular Weight (g/mol) 177.20
PubChem CID 712398
Formula Weight 177.2
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material D-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • (R)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.