Learn More
Thermo Scientific™ D.A.R.T.'S™ (Disposable Automation Research Tips), with Extended Length
GREENER_CHOICE
Achieve ultimate performance in automated pipetting with Thermo Scientific™ D.A.R.T.'S™ Extended Length tips — superior quality with the flexibility to choose tips for any application.
Brand:  Thermo Scientific™ 5562-11
240.99 EUR valid until 2025-12-31
Use promo code "24288" to get your promotional price.
 View more versions of this product
Product Code. 10621354
  • 269.00 € / Pack of 1920
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Perform pipetting more effectively with Thermo Scientific™ Matrix™ SerialMate™ Extended Length Pipette Tips, for use with the Matrix SerialMate™ Pipetter. Unlike conventional automation tips, D.A.R.T.'s™ are packaged in disposable rigid magazines and seal onto the pipetting heads by being pressed against a silicone pad. This definitive seal minimizes volume displacement, ensures tip straightness and enables consistent dispensing for improved well access. A small internal orifice provides small droplet formation resulting in optimal accuracy and precision.
Features:
  • Stepped-body design minimizes volume displacement and ensures tip straightness for better well access.
  • Highly polished molds and cores ensure low liquid retention.
  • Small internal tip orifice provides better small volume droplet formation
  • Sterile, sterile filtered, wide-bore, and extended-length tip options are available
  • Tips are thoroughly inspected for proper formation and straightness
  • Unique face seal design ensures consistent tip-to-tip height for precise low volume dispensing.
  • Entire assembly can be quickly removed and replaced eliminating extra handling of the tips, which can cause damage or contamination.
Includes: 20 magazines per case
Compatible with: Thermo Scientific™ Matrix™ Hydra™ DT, Matrix™ PlateMate™ 2x3 and PlateMate™ Plus Systems.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
220μl DeepWell Sterile Pipette Tips
Filtered
3-10%
ISO 9001 and ISO 13485
Automation
Thermo Scientific Matrix SerialMate
220 μL
Sterile
3-10%
Free of RNase, DNase, DNA, ATP, and endotoxin
Single-use
D.A.R.T.S