missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ D.A.R.T.'S™ (Disposable Automation Research Tips)
GREENER_CHOICE

Product Code. 10171393
Click to view available options
Volume (Metric):
12.5 μL
30 μL
100 μL
220 μL
250 μL
Sterility:
Non-sterile
Sterile
Packaging:
10 x 384 tips
20 x 384 tips
20 x 96 tips
Unit Size:
Pack of 1920
Pack of 3840
Pack of 7680
Filtered:
Filtered
Non-filtered
This item is not returnable. View return policy

Product Code. 10171393

Brand: Thermo Scientific™ 5322

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Achieve ultimate performance in automated pipetting with these tips--superior quality with the flexibility to choose tips for any application.

Perform pipetting more effectively with Thermo Scientific™ Matrix™ SerialMate™ Pipette Tips, for use with the Matrix SerialMate™ Pipetter. Unlike conventional automation tips, D.A.R.T.'s™ are packaged in disposable rigid magazines and seal onto the pipetting heads by being pressed against a silicone pad. This definitive seal minimizes volume displacement, ensures tip straightness and enables consistent dispensing for improved well access. A small internal orifice provides small droplet formation resulting in optimal accuracy and precision.

Features:
  • Stepped-body design minimizes volume displacement and ensures tip straightness for better well access.
  • Highly polished molds and cores ensure low liquid retention.
  • Small internal tip orifice provides better small volume droplet formation
  • Sterile, sterile filtered, wide-bore, and extended-length tip options are available
  • Tips are thoroughly inspected for proper formation and straightness
  • Unique face seal design ensures consistent tip-to-tip height for precise low volume dispensing.
  • Entire assembly can be quickly removed and replaced eliminating extra handling of the tips, which can cause damage or contamination.

Includes: 20 magazines per case

Compatible with: Thermo Scientific™ Matrix™ Hydra™ DT, Matrix™ PlateMate™ 2x3 and PlateMate™ Plus Systems.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 100 μL
Sterility Sterile
Color Natural
Autoclavable Not Autoclavable
Certifications/Compliance ISO 9001 and ISO 13485
Clean Claims Free of RNase, DNase, DNA, ATP, and endotoxin
Format Rack
Tip Style Automation
Packaging 10 x 384 tips
Quantity 3840 tips
Disposable Single-use
For Use With (Equipment) Thermo Scientific Matrix SerialMate
Product Line D.A.R.T.S
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.