missing translation for 'onlineSavingsMsg'
Learn More

D-AP5, Tocris Bioscience™

Product Code. 18004362
Click to view available options
Quantity:
1 mg
10 mg
50 mg
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective NMDA receptor antagonist; more active form of DL-AP5

Widely used competitive NMDA receptor antagonist. More active form of AP5. Also agonist at quisqualate-sensitized AP6 site where it is less potent than the L-isomer, L-AP5 (Cat. No.0107).

DL Mixture and L-isomer also available.

Chemical Identifiers

CAS 79055-68-8
Molecular Formula C5H12NO5P
Molecular Weight (g/mol) 197.127
InChI Key VOROEQBFPPIACJ-SCSAIBSYSA-N
Synonym d-ap5, 5-phosphono-d-norvaline, d-norvaline, 5-phosphono, d-apv, 2r-2-amino-5-phosphonopentanoic acid, d-2-amino-5-phosphopentanoic acid, d---2-amino-5-phosphonopentanoic acid, d--apv, d-2-amino-5-phosphonovaleric acid, d--ap-5
PubChem CID 135342
IUPAC Name (2R)-2-amino-5-phosphonopentanoic acid
SMILES C(CC(C(=O)O)N)CP(=O)(O)O

Specifications

Quantity 10 mg
Formula Weight 197.13
Percent Purity >99%
Chemical Name or Material D-AP5
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.