missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Arginine monohydrochloride, 98%

Code produit. 11413327
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
25 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

The influence in creatinine metabolismis that by that d-arginine monohydrochloride caused slightly larger increases in muscle creatine. D-Arginyl ethyl ester was synthesized by vigorously stirring D-arginine monohydrochloride and excess anhydrous ethanol in an ice bath.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The influence in creatinine metabolismis that by that d-arginine monohydrochloride caused slightly larger increases in muscle creatine. D-Arginyl ethyl ester was synthesized by vigorously stirring D-arginine monohydrochloride and excess anhydrous ethanol in an ice bath.

Solubility
Soluble in water (50 mg/ml), and 3N HCl (5).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 627-75-8
Molecular Formula C6H15ClN4O2
Molecular Weight (g/mol) 210.662
MDL Number MFCD00012620
InChI Key KWTQSFXGGICVPE-PGMHMLKASA-N
Synonym d-arginine hydrochloride, h-d-arg-oh.hcl, d-arginine monohydrochloride, d-arginine, monohydrochloride, unii-aw80ygd17d, d-arginine hcl, arginine, monohydrochloride, d, aw80ygd17d, arginine, chloride, d--arginine hydrochloride
PubChem CID 12326
IUPAC Name (2R)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN=C(N)N.Cl

Spécification

Melting Point 216°C to 218°C
Beilstein 5773432
Solubility Information Soluble in water (50mg/ml),and 3N HCl (5).
Formula Weight 210.67
Percent Purity 99%
Quantity 1 g
Sensitivity Hygroscopic
Chemical Name or Material D-Arginine monohydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'