missing translation for 'onlineSavingsMsg'
Learn More
D-Asparagine monhydrate, 100.5%, MP Biomedicals™ Product Code.: 11313979

D-Asparagine monhydrate, 100.5%, MP Biomedicals™

Product Code. 11313979 Shop All MP Biomedicals Products
1 kg
Click to view available options
Quantity:
1 kg
This item is not returnable. View return policy

Product Code. 11313979

Brand: MP Biomedicals 0210079491

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 5794-24-1
Molecular Formula C4H10N2O4
Molecular Weight (g/mol) 150.134
InChI Key RBMGJIZCEWRQES-HSHFZTNMSA-N
Synonym d-asparagine monohydrate, d-asparagine hydrate, r-2,4-diamino-4-oxobutanoic acid hydrate, d-asparagine, monohydrate, d-asparaginemonohydrate, d---asparagine monohydrate, d-aspartic acid 4-amide, r---2-aminosuccinamic acid, d--asparagine monohydrate, r-2-aminosuccinic acid 4-amide
PubChem CID 16211197
IUPAC Name (2R)-2,4-diamino-4-oxobutanoic acid;hydrate
SMILES C(C(C(=O)O)N)C(=O)N.O

Specifications

Assay Percent Range 1.005
Quantity 1 kg
Formula Weight 150.1
Physical Form Crystalline
Chemical Name or Material D-Asparagine monhydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
D-Asparagine monhydrate, 100.5%, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.