missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Asparagine monohydrate

Product Code. 17271136
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5794-24-1
Molecular Weight (g/mol) 150.13
InChI Key RBMGJIZCEWRQES-UHFFFAOYNA-N
IUPAC Name 2-amino-3-carbamoylpropanoic acid hydrate
SMILES O.NC(CC(N)=O)C(O)=O

Specifications

Melting Point 280.0°C
Molecular Formula C4H10N2O4
Linear Formula H2NCOCH2CH(NH2)CO2H·H2O
Beilstein 04, 471
Merck Index 14, 837
Solubility Information Solubility aq. soln.: 35 g/L (20°C). Other solubilities: soluble in acids and alkalies, practically insoluble in methanol, ethanol, ether, and benzene
Quantity 1 g
Formula Weight 150.13
Chemical Name or Material D-Asparagine monohydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'