missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-(-)-Asparagine monohydrate, 99%
Click to view available options
Quantity:
25 g
100 g
Description
D-(-)-Asparagine monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-(-)-Asparagine monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Soluble in water (35 g/L at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
D-(-)-Asparagine monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Soluble in water (35 g/L at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Specifications
Specifications
Melting Point | 274°C to 276°C (decomposition) |
Merck Index | 14,837 |
Solubility Information | Soluble in water (35g/L at 20°C). |
Quantity | 100 g |
Formula Weight | 150.14 (132.12 Anhydrous) |
Percent Purity | 99% |
Chemical Name or Material | D-(-)-Asparagine monohydrate |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific Chemicals D-(-)-Asparagine monohydrate, 99% >
Spot an opportunity for improvement?Share a Content Correction