missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Cysteine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
D-Cysteine is an important source of sulfide in human metabolism. It is the precursor for glutathione. It plays important role in protein structure and in metal binding.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-Cysteine is an important source of sulfide in human metabolism. It is the precursor for glutathione. It plays important role in protein structure and in metal binding.
Solubility
Soluble in water (partly).
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
D-Cysteine is an important source of sulfide in human metabolism. It is the precursor for glutathione. It plays important role in protein structure and in metal binding.
Solubility
Soluble in water (partly).
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Specifications
Melting Point | 230°C (decomposition) |
Beilstein | 1721407 |
Solubility Information | Soluble in water (partly). |
Quantity | 1 g |
Formula Weight | 121.16 |
Sensitivity | Air Sensitive |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | D-Cysteine |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction