missing translation for 'onlineSavingsMsg'
Learn More
D-Cysteine, 98%, Thermo Scientific Chemicals Product Code.: 15449138

D-Cysteine, 98%, Thermo Scientific Chemicals

Product Code. 15449138
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15449138

Brand: Thermo Scientific Alfa Aesar H62627.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

D-Cysteine is an important source of sulfide in human metabolism. It is the precursor for glutathione. It plays important role in protein structure and in metal binding.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Cysteine is an important source of sulfide in human metabolism. It is the precursor for glutathione. It plays important role in protein structure and in metal binding.

Solubility
Soluble in water (partly).

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 921-01-7
Molecular Formula C3H7NO2S
Molecular Weight (g/mol) 121.15
MDL Number MFCD00066461
InChI Key XUJNEKJLAYXESH-RUOZJJLTNA-N
Synonym d-cysteine, d-cystein, 2s-2-amino-3-sulfanylpropanoic acid, d-zystein, cysteine, d, s-2-amino-3-mercaptopropanoic acid, d-amino-3-mercaptopropionic acid, d-cys, h-d-cys-oh, cysteine
PubChem CID 92851
ChEBI CHEBI:16375
IUPAC Name (2S)-2-amino-3-sulfanylpropanoic acid
SMILES N[C@H](CS)C(O)=O

Specifications

Melting Point 230°C (decomposition)
Beilstein 1721407
Solubility Information Soluble in water (partly).
Quantity 5 g
Formula Weight 121.16
Sensitivity Air Sensitive
Percent Purity 98%
Physical Form Solid
Chemical Name or Material D-Cysteine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
D-Cysteine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.