missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals D-Cysteine hydrochloride monohydrate, 99% Product Code.: 11416200

Thermo Scientific Chemicals D-Cysteine hydrochloride monohydrate, 99%

Product Code. 11416200
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11416200

Brand: Thermo Scientific Chemicals H27107.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207121-46-8
Molecular Formula C3H10ClNO3S
Molecular Weight (g/mol) 175.627
MDL Number MFCD00150051
InChI Key QIJRTFXNRTXDIP-YBBRRFGFSA-N
Synonym d-cysteine hydrochloride monohydrate, s-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, d-cysteine*hcl*h2o, d-cysteine hydrochloride hydrate, h-d-cys-oh.hcl.h2o, d-cysteine hydrate hydrochloride, c3h7no2s.hcl.h2o, d-cysteine hcl monohydrate, pubchem12314, h-d-cys-oh??hcl??h2o
PubChem CID 10176341
IUPAC Name (2S)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
SMILES C(C(C(=O)O)N)S.O.Cl

Specifications

Melting Point 184°C to 186°C
Quantity 5 g
Formula Weight 175.64 (157.62 Anhydrous)
Optical Rotation −5.5° (c=5 in 5N HCl)
Percent Purity 99%
Chemical Name or Material D-Cysteine hydrochloride monohydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10248240 Certificate TypeCertificate of Analysis Date19/10/2023 Catalog NumberH27107.06,H27107.03
Lot Number10203787 Certificate TypeCertificate of Analysis Date15/09/2023 Catalog NumberH27107.06,H27107.03
Lot Number10212593 Certificate TypeCertificate of Analysis Date15/09/2023 Catalog NumberH27107.06,H27107.03
Lot Number10203787 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27107.06,H27107.03
Lot Number10174289 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27107.06,H27107.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals D-Cysteine hydrochloride monohydrate, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.