Learn More
IBA Lifesciences Desthiobiotin
GREENER_CHOICE
Desthiobiotin powder for preparation of Strep-Tactin™ elution buffer
260.00 € - 1125.00 €
Specifications
For Use With (Application) For Elution of Strep-Tag II Proteins From Strep-Tactin
Product Type Desthiobiotin
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12753064
GREENER_CHOICE
View Documents
IBA Lifesciences
2-1000-002
1 g
260.00 €
Each
12763064
GREENER_CHOICE
View Documents
IBA Lifesciences
2-1000-005
5 g
1125.00 €
Each
Description
Description
Desthiobiotin is a modified form of biotin that displaces Strep-tag™II and Twin-Strep-tag™ fusion proteins from the modified biotin-binding pocket of Strep-Tactin™ in a competitive manner, resulting in a mild elution.
Desthiobiotin powder can be used for preparation of Strep-Tactin™ elution buffer, Buffer E.
Please note that desthiobiotin is not applicable for elution from Strep-Tactin™XT resins.
Specifications
Specifications
For Elution of Strep-Tag II Proteins From Strep-Tactin
Desthiobiotin