missing translation for 'onlineSavingsMsg'
Learn More

D-Epoxone, 98%, Thermo Scientific Chemicals

49.95 € - 194.97 €

Chemical Identifiers

CAS 18422-53-2
Molecular Formula C12H18O6
Molecular Weight (g/mol) 258.27
MDL Number MFCD00063383
InChI Key IVWWFWFVSWOTLP-RDNCKJRISA-N
Synonym 3ar,4'r,7ar-2,2,2',2'-tetramethyldihydrospiro 1,3 dioxolo 4,5-c pyran-6,4'-1,3 dioxolan-7 7ah-one
PubChem CID 118990039
IUPAC Name (4S,7'aR)-2,2,2',2'-tetramethylspiro[1,3-dioxolane-4,6'-4,7a-dihydro-3aH-[1,3]dioxolo[4,5-c]pyran]-7'-one
SMILES CC1(OCC2(O1)C(=O)C3C(CO2)OC(O3)(C)C)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11389565
SDS View Documents
Thermo Scientific Alfa Aesar
L18989.03
1 g
49.95 €
1g
Please to purchase this item. Need a web account? Register with us today!
11399565
SDS View Documents
Thermo Scientific Alfa Aesar
L18989.06
5 g
194.97 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

D-Epoxone is a chemical reagent used for enantioselective epoxidation of alkenes. It is involved in the synthesis of Irciniastatin (A and B), which are used as antitumor compounds. It is used as cytotoxic secondary metabolite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Epoxone is a chemical reagent used for enantioselective epoxidation of alkenes. It is involved in the synthesis of Irciniastatin (A and B), which are used as antitumor compounds. It is used as cytotoxic secondary metabolite.

Solubility
Soluble in chloroform and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

18422-53-2
258.27
IVWWFWFVSWOTLP-RDNCKJRISA-N
118990039
CC1(OCC2(O1)C(=O)C3C(CO2)OC(O3)(C)C)C
C12H18O6
MFCD00063383
3ar,4'r,7ar-2,2,2',2'-tetramethyldihydrospiro 1,3 dioxolo 4,5-c pyran-6,4'-1,3 dioxolan-7 7ah-one
(4S,7'aR)-2,2,2',2'-tetramethylspiro[1,3-dioxolane-4,6'-4,7a-dihydro-3aH-[1,3]dioxolo[4,5-c]pyran]-7'-one

Specifications

18422-53-2
C12H18O6
1319073
Soluble in chloroform and methanol.
CC1(OCC2(O1)C(=O)C3C(CO2)OC(O3)(C)C)C
258.27
258.27
D-Epoxone
101°C to 103°C
MFCD00063383
3ar,4'r,7ar-2,2,2',2'-tetramethyldihydrospiro 1,3 dioxolo 4,5-c pyran-6,4'-1,3 dioxolan-7 7ah-one
IVWWFWFVSWOTLP-RDNCKJRISA-N
(4S,7'aR)-2,2,2',2'-tetramethylspiro[1,3-dioxolane-4,6'-4,7a-dihydro-3aH-[1,3]dioxolo[4,5-c]pyran]-7'-one
118990039
98%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.