missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford D-Fructose
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
58.15 € - 81.10 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30018276
|
Melford
F21500-1000.0 |
1 kg |
81.10 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30018277
|
Melford
F21500-500.0 |
500 g |
58.15 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- Substance Name: Levulose
- CAS Number: 57-48-7
- Molecular Formula: C6H12O6
- Molecular Weight: 180.16
Specifications
57-48-7 | |
C6H12O6 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Melford D-Fructose